2-Amino-4-phenylpyrimidine ≥97%
Supplier: Aladdin Scientific
|
Warning
|
A168818-1G
A168818-200MG
A168818-5G
ALA168818-1GEA
49.31
USD
ALA168818-1G
ALA168818200MG
ALA168818-5G
2-Amino-4-phenylpyrimidine ≥97%
2-Amino-4-phenylpyrimidine
Formula:
C₁₀H₉N₃ Melting Pt: 162-164°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00094706 CAS Number: 2305-87-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...