2-Aminomethyl-1H-imidazole dihydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Aminomethyl-1H-imidazole dihydrochloride, 2-(Aminomethyl)imidazole dihydrochloride, (1H-Imidazol-2-yl)methanamine dihydrochloride

A168767-1G A168767-5G
ALA168767-1GEA 66.41 USD
ALA168767-1G ALA168767-5G
2-Aminomethyl-1H-imidazole dihydrochloride ≥97%
2-Aminomethyl-1H-imidazole dihydrochloride
Formula: C₄H₉Cl₂N₃
Melting Pt: 234-241 °C
Storage Temperature: Ambient
MDL Number: MFCD06738779
CAS Number: 22600-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5469bfa4a25c74f33f71274b005bd4750955679&vid=c5469bfa4a25c74f33f71274b005bd4750955679&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR