(2-Amino-2-oxoethoxy)acetic acid ≥95%

Supplier: ALADDIN SCIENTIFIC

A168692-1G A168692-250MG A168692-50MG
ALA168692-1GEA 284.91 USD
ALA168692-1G ALA168692250MG ALA168692-50MG
(2-Amino-2-oxoethoxy)acetic acid ≥95%
(2-Amino-2-oxoethoxy)acetic acid
Formula: C₄H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00176328
CAS Number: 22064-40-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c476712ddd4587624869c6d202c0961c381e783e&vid=c476712ddd4587624869c6d202c0961c381e783e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR