ANA-12 ≥96%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
N-(2-((2-oxoazepan-3-yl)carbamoyl)phenyl)benzo[b]thiophene-2-carboxamide, N-[2-[[(Hexahydro-2-oxo-1H-azepin-3-yl)amino]carbonyl]phenyl]benzo[b]thiophene-2-carboxamide, N-[2-[(2-oxoazepan-3-yl)carbamoyl]phenyl]-1-benzothiophene-2-carboxamide
A168673-100MG
A168673-10MG
A168673-25MG
A168673-50MG
A168673-5MG
ALA168673100MGEA
761.81
USD
ALA168673100MG
ALA168673-10MG
ALA168673-25MG
ALA168673-50MG
ALA168673-5MG
ANA-12 ≥96%
ANA-12
ANA-12 is an effective and selective TrkB antagonist with IC50 of 45.6 nM and 41.1 μM for high affinity and low affinity sites respectively.
Formula:
C₂₂H₂₁N₃O₃S MW: 407.49 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00117444 CAS Number: 219766-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...