3-Aminobutan-2-one hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

A168580-250MG A168580-100MG A168580-1G
ALA168580250MGEA 120.56 USD
ALA168580250MG ALA168580100MG ALA168580-1G
3-Aminobutan-2-one hydrochloride ≥95%
3-Aminobutan-2-one hydrochloride
Formula: C₄H₁₀ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD16619121
CAS Number: 21419-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=909e3ac3414a78942496edacab35654d5bb53580&vid=909e3ac3414a78942496edacab35654d5bb53580&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR