1-Acetylpiperidin-4-amine hydrochloride ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-Acetyl-4-aminopiperidine hydrochloride, 1-(4-Aminopiperidin-1-yl)ethanone hydrochloride

A168578-1G A168578-250MG
ALA168578-1GEA 36.01 USD
ALA168578-1G ALA168578250MG
1-Acetylpiperidin-4-amine hydrochloride ≥96%
1-Acetylpiperidin-4-amine hydrochloride
Formula: C₇H₁₅ClN₂O
MW: 178.6600 g/mol
Melting Pt: 240-245 °C
Storage Temperature: Ambient
MDL Number: MFCD08690151
CAS Number: 214147-48-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e19cb083e2fec8d6a2c73571006c72c7522f48ea&vid=e19cb083e2fec8d6a2c73571006c72c7522f48ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR