4-Amino-2-chlorobenzamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A168543-1G A168543-250MG
ALA168543-1GEA 201.31 USD
ALA168543-1G ALA168543250MG
4-Amino-2-chlorobenzamide ≥97%
4-Amino-2-chlorobenzamide
Formula: C₇H₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09043062
CAS Number: 211374-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f65dbe6fb0ca52a8ab008aaa7a63b95ba74276aa&vid=f65dbe6fb0ca52a8ab008aaa7a63b95ba74276aa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR