2-Amino-5-bromobenzylalkohol ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A168462-1G A168462-250MG A168462-25G A168462-5G
ALA168462-1GEA 26.51 USD
ALA168462-1G ALA168462250MG ALA168462-25G ALA168462-5G
2-Amino-5-bromobenzylalkohol ≥95%
2-Amino-5-bromobenzylalkohol
Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD08234509
CAS Number: 20712-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c26ec58ec4877305a491cc4914830744f6eeb5f&vid=1c26ec58ec4877305a491cc4914830744f6eeb5f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR