Allyl crotonate ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A168419-1G A168419-250MG A168419-25G A168419-5G
ALA168419-1GEA 65.46 USD
ALA168419-1G ALA168419250MG ALA168419-25G ALA168419-5G
Allyl crotonate ≥97%
Allyl crotonate
Formula: CH₃CH=CHCO₂CH₂CH=CH₂
MW: 126.157 g/mol
Boiling Pt: 63-65℃
Density: 0.944 g/cm³
Flash Pt: 48°C
Storage Temperature: Ambient
MDL Number: MFCD00026990
CAS Number: 20474-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d15c4e080de335201c41b0868cf87530d555f454&vid=d15c4e080de335201c41b0868cf87530d555f454&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR