(2-Aminoethyl)phosphonic acid ≥99%

Supplier: Aladdin Scientific
Warning

A168405-1G A168405-250MG A168405-25G A168405-5G
ALA168405-1GEA 179.46 USD
ALA168405-1G ALA168405250MG ALA168405-25G ALA168405-5G
(2-Aminoethyl)phosphonic acid ≥99%
(2-Aminoethyl)phosphonic acid

The surface of gold nanoparticles (Au NPs) were functionalized with 2-aminoethylphosphonic acid that exhibited calcium affinity which enabled targeted delivery of Au NPs to calcified tissue.2-Aminoethylphosphonic acid was used as a growth medium for the marine bacterium Roseovarius nubinhibens ISM.

Formula: C₂H₈NO₃P
Melting Pt: 296°C
Storage Temperature: Ambient
MDL Number: MFCD00008182
CAS Number: 2041-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c39f61a3cf3596847a8a6dd2f5092612556a3b6a&vid=c39f61a3cf3596847a8a6dd2f5092612556a3b6a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR