2-Acetamido-5-chlorothiazole ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: N-(5-Chlorothiazol-2-yl)acetamide

A168368-10G A168368-1G A168368-5G
ALA168368-10GEA 63.56 USD
ALA168368-10G ALA168368-1G ALA168368-5G
2-Acetamido-5-chlorothiazole ≥96%
2-Acetamido-5-chlorothiazole
Formula: C₅H₅ClN₂OS
MW: 176.63 g/mol
Melting Pt: 208-212°C
Storage Temperature: Ambient
MDL Number: MFCD01318098
CAS Number: 20256-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=357395ad627bf837da416da21d590e753607963c&vid=357395ad627bf837da416da21d590e753607963c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR