tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate ≥97%

Supplier: Aladdin Scientific
Warning

A168231-1G A168231-5G
ALA168231-1GEA 55.96 USD
ALA168231-1G ALA168231-5G
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate ≥97%
tert-Butyl-4-(2-aminoethyl)piperazine-1-carboxylate
Formula: C₁₁H₂₃N₃O₂
Flash Pt: >198 °C
Storage Temperature: Ambient
MDL Number: MFCD02683049
CAS Number: 192130-34-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d8b33a89702d7499981b1812bce8154c5388de6&vid=5d8b33a89702d7499981b1812bce8154c5388de6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR