Methyl 3-amino-5-methylbenzoate ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Methyl 3-amino-5-methylbenzoate, 3-Amino-5-methyl-benzoic acid methyl ester

A168134-10G A168134-1G A168134-250MG A168134-5G
ALA168134-10GEA 448.31 USD
ALA168134-10G ALA168134-1G ALA168134250MG ALA168134-5G
Methyl 3-amino-5-methylbenzoate ≥97%
Methyl 3-amino-5-methylbenzoate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263222
CAS Number: 18595-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb2928dfc0de75402bcfc7ae42d91e3fe5b67544&vid=bb2928dfc0de75402bcfc7ae42d91e3fe5b67544&prid=web&clid=SAPDAM
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR