4-Azetidin-3-yl-thiomorpholine-1,1-dioxide dihydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A167988-1G A167988-250MG A167988-50MG
ALA167988-1GEA 1101.91 USD
ALA167988-1G ALA167988250MG ALA167988-50MG
4-Azetidin-3-yl-thiomorpholine-1,1-dioxide dihydrochloride ≥97%
4-Azetidin-3-yl-thiomorpholine-1,1-dioxide dihydrochloride
Formula: C₇H₁₄N₂O₂S·2ClH
MW: 263.185 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22123267
CAS Number: 178312-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea624161614f877640e41008d4d87eeaf5bc0d5&vid=bea624161614f877640e41008d4d87eeaf5bc0d5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR