5-Amino-1-(3-chloro-4-fluorophenyl)-1H-pyrazole-4-carbonitrile 97

Supplier: ALADDIN SCIENTIFIC

A167931-1G A167931-250MG A167931-50MG
ALA167931-1GEA 488.21 USD
ALA167931-1G ALA167931250MG ALA167931-50MG
5-Amino-1-(3-chloro-4-fluorophenyl)-1H-pyrazole-4-carbonitrile 97
5-Amino-1-(3-chloro-4-fluorophenyl)-1H-pyrazole-4-carbonitrile
Formula: C₁₀H₆ClFN₄
MW: 236.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052032
CAS Number: 175135-53-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=210c78bdccd0ae4313f6f73e243faf4a6f685b17&vid=210c78bdccd0ae4313f6f73e243faf4a6f685b17&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR