4-Amino-1,8-naphthalimide ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A167915-1G
A167915-250MG
A167915-50MG
A167915-5G
ALA167915-1GEA
182.31
USD
ALA167915-1G
ALA167915250MG
ALA167915-50MG
ALA167915-5G
4-Amino-1,8-naphthalimide ≥95%
4-Amino-1,8-naphthalimide
Formula:
C₁₂H₈N₂O₂ Melting Pt: >360°C Density: 1.105 g/cm³ Storage Temperature: Freezer |
MDL Number:
MFCD00006921 CAS Number: 1742-95-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...