3-(2-Pyridyl)-DL-alanine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Amino-3-(pyridin-2-yl)propionic acid, H-DL-β-(2-Pyridyl)-Ala-OH, H-β-(2-Pyridyl)-DL-Ala-OH
A167909-100MG
A167909-1G
A167909-250MG
A167909-5G
ALA167909100MGEA
52.16
USD
ALA167909100MG
ALA167909-1G
ALA167909250MG
ALA167909-5G
3-(2-Pyridyl)-DL-alanine ≥98%
3-(2-Pyridyl)-DL-alanine
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00237828 CAS Number: 17407-44-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...