3-Amino-2-bromobenzoic acid ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A167825-1G
A167825-250MG
A167825-5G
ALA167825-1GEA
44.56
USD
ALA167825-1G
ALA167825250MG
ALA167825-5G
3-Amino-2-bromobenzoic acid ≥95%
3-Amino-2-bromobenzoic acid
Formula:
C₇H₆BrNO₂ MW: 216.03 Melting Pt: 153-158 °C Storage Temperature: Ambient |
MDL Number:
MFCD07786957 CAS Number: 168899-61-4 UN: 2811 ADR: 6.1 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...