4-(2-Aminoethyl)benzonitrile hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A167799-1G A167799-250MG A167799-5G
ALA167799-1GEA 56.91 USD
ALA167799-1G ALA167799250MG ALA167799-5G
4-(2-Aminoethyl)benzonitrile hydrochloride ≥97%
4-(2-Aminoethyl)benzonitrile hydrochloride
Formula: C₉H₁₁ClN₂
Melting Pt: 211-216 °C
CAS Number: 167762-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b42b6e55bb37d1ed3fa2e856960cec0bcc7f7dac&vid=b42b6e55bb37d1ed3fa2e856960cec0bcc7f7dac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR