2-Chloro-3-amino-4-(trifluoromethyl)pyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Amino-2-chloro-4-(trifluoromethyl)pyridine

A167784-1G A167784-250MG A167784-5G
ALA167784-1GEA 421.71 USD
ALA167784-1G ALA167784250MG ALA167784-5G
2-Chloro-3-amino-4-(trifluoromethyl)pyridine ≥97%
2-Chloro-3-amino-4-(trifluoromethyl)pyridine
Formula: C₆H₄ClF₃N₂
Melting Pt: 53-59 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09863900
CAS Number: 166770-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be5e4c62f6d544bf8c773aa7096a0b8bbdc71648&vid=be5e4c62f6d544bf8c773aa7096a0b8bbdc71648&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR