3-Aminomethyltetrahydrofuran ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: (Tetrahydrofuran-3-yl)methanamine, 3-(Aminomethyl)oxolane, (tetrahydrofuran-3-yl)methanamine, Oxolan-3-ylmethanamine, 3-(Aminomethyl)tetrahydrofuran

A167762-1G A167762-250MG A167762-25G A167762-5G
ALA167762-1GEA 30.31 USD
ALA167762-1G ALA167762250MG ALA167762-25G ALA167762-5G
3-Aminomethyltetrahydrofuran ≥95%
3-Aminomethyltetrahydrofuran
Formula: C₅H₁₁NO
Boiling Pt: 67 °C
Density: 1 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD08234925
CAS Number: 165253-31-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8d31487465d181eaad573fdef441bf31d33d69f&vid=c8d31487465d181eaad573fdef441bf31d33d69f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR