10-Acetylphenothiazine ≥98%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
1-(10H-Phenothiazin-10-yl)ethanone, 10-Acetyl-10H-phenothiazine
A167717-1G
A167717-250MG
A167717-25G
A167717-5G
ALA167717-1GEA
108.21
USD
ALA167717-1G
ALA167717250MG
ALA167717-25G
ALA167717-5G
10-Acetylphenothiazine ≥98%
10-Acetylphenothiazine
Formula:
C₁₄H₁₁NOS Melting Pt: 204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022264 CAS Number: 1628-29-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...