5-Phenyl-1,3,4-oxadiazol-2-amine ≥97%

Supplier: Aladdin Scientific
Warning

A167683-10G A167683-1G A167683-250MG A167683-5G
ALA167683-10GEA 200.36 USD
ALA167683-10G ALA167683-1G ALA167683250MG ALA167683-5G
5-Phenyl-1,3,4-oxadiazol-2-amine ≥97%
5-Phenyl-1,3,4-oxadiazol-2-amine
Formula: C₈H₇N₃O
Melting Pt: 237-242°C
Storage Temperature: Ambient
MDL Number: MFCD00126383
CAS Number: 1612-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcfd40b271fbd0baed852c5455a4d35dd2c0f60b&vid=bcfd40b271fbd0baed852c5455a4d35dd2c0f60b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR