2-Amino-4-(trifluoromethyl)pyrimidine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A167675-100G A167675-1G
ALA167675-100GEA 1842.91 USD
ALA167675-100G ALA167675-1G
2-Amino-4-(trifluoromethyl)pyrimidine ≥97%
2-Amino-4-(trifluoromethyl)pyrimidine
Formula: C₅H₄F₃N₃
MW: 163.1 g/mol
Melting Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00828624
CAS Number: 16075-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90fa91c77d2ae8c1533f260aa51fa0c81f92264c&vid=90fa91c77d2ae8c1533f260aa51fa0c81f92264c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR