1-Allyl-2-methylbenzene ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A167621-1G A167621-25G A167621-5G
ALA167621-1GEA 39.81 USD
ALA167621-1G ALA167621-25G ALA167621-5G
1-Allyl-2-methylbenzene ≥97%
1-Allyl-2-methylbenzene
Formula: C₁₀H₁₂
Density: 0.895 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00060911
CAS Number: 1587-04-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b89f77dc6cf05a69887bf0ea510c8c01138555f&vid=4b89f77dc6cf05a69887bf0ea510c8c01138555f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR