Triapine ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone

A167286-25MG A167286-5MG A167286-100MG A167286-10MG
ALA167286-25MGEA 179.46 USD
ALA167286-25MG ALA167286-5MG ALA167286100MG ALA167286-10MG
Triapine ≥98% (by HPLC)
Triapine
Formula: C₇H₉N₅S
MW: 195.24 g/mol
Melting Pt: 234 °C
Storage Temperature: Freezer
MDL Number: MFCD00924747
CAS Number: 143621-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39fc907c7c6a898405a19f203bb1231fff55d9b2&vid=39fc907c7c6a898405a19f203bb1231fff55d9b2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR