2-Acetylamino-5-bromo-3-methylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A167254-1G A167254-250MG A167254-5G
ALA167254-1GEA 46.46 USD
ALA167254-1G ALA167254250MG ALA167254-5G
2-Acetylamino-5-bromo-3-methylpyridine ≥97%
2-Acetylamino-5-bromo-3-methylpyridine
Formula: C₈H₉BrN₂O
MW: 229.07 g/mol
Melting Pt: 111-115 °C
Storage Temperature: Ambient
MDL Number: MFCD01928227
CAS Number: 142404-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=541a4ce0cda015d32a7419702ed3721b78fee7fb&vid=541a4ce0cda015d32a7419702ed3721b78fee7fb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR