7-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
7-Amino-4-methyl-2H-benzo[b][1,4]oxazin-3(4H)-one
A167206-100MG
A167206-1G
A167206-250MG
A167206-50MG
ALA167206100MGEA
39.81
USD
ALA167206100MG
ALA167206-1G
ALA167206250MG
ALA167206-50MG
7-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥97%
7-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one
Formula:
C₉H₁₀N₂O₂ MW: 178.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659594 CAS Number: 141068-81-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...