1-Allylpiperazine ≥96%

Supplier: ALADDIN SCIENTIFIC
Danger

A167179-1G A167179-25G
ALA167179-1GEA 17.01 USD
ALA167179-1G ALA167179-25G
1-Allylpiperazine ≥96%
1-Allylpiperazine
Formula: C₇H₁₄N₂
Boiling Pt: 181℃
Density: 0.902 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00167970
CAS Number: 13961-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=188cd7476d9269e65ee78a85b799264225b651dc&vid=188cd7476d9269e65ee78a85b799264225b651dc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR