4-Aminobenzoate potassium salt ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A167159-100G A167159-25G A167159-500G A167159-5G
ALA167159-100GEA 60.71 USD
ALA167159-100G ALA167159-25G ALA167159-500G ALA167159-5G
4-Aminobenzoate potassium salt ≥97%
4-Aminobenzoate potassium salt
Formula: C₇H₆KNO₂
Storage Temperature: Ambient
MDL Number: MFCD00064911
CAS Number: 138-84-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e075d3bb89412ee26271aa925640817668bc370&vid=0e075d3bb89412ee26271aa925640817668bc370&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR