2-Amino-6-chloro-3-methylquinoline ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

A167119-1G A167119-250MG
ALA167119-1GEA 201.31 USD
ALA167119-1G ALA167119250MG
2-Amino-6-chloro-3-methylquinoline ≥95%
2-Amino-6-chloro-3-methylquinoline
Formula: C₁₀H₉ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00272509
CAS Number: 137110-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e0333d5e16916af87dbb597da7eb007d2329b99&vid=9e0333d5e16916af87dbb597da7eb007d2329b99&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR