1-(4-Aminophenyl)pyrrolidin-2-one ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(4-Aminophenyl)pyrrolidin-2-one, 1-(4-Aminophenyl)-2-pyrrolidone, 1-(4-Aminophenyl)-2-pyrrolidinone

A167110-1G A167110-250MG A167110-5G
ALA167110-1GEA 233.61 USD
ALA167110-1G ALA167110250MG ALA167110-5G
1-(4-Aminophenyl)pyrrolidin-2-one ≥98%
1-(4-Aminophenyl)pyrrolidin-2-one
Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100993
CAS Number: 13691-22-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ba628927bf0c4170c73c1703f1619713a848d80&vid=2ba628927bf0c4170c73c1703f1619713a848d80&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR