2-Acetyl-4-methylthiophene ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-(4-Methyl-2-thienyl)-1-ethanone, 1-(4-Methylthiophen-2-yl)ethanone

A167104-1G A167104-25G A167104-5G
ALA167104-1GEA 34.11 USD
ALA167104-1G ALA167104-25G ALA167104-5G
2-Acetyl-4-methylthiophene ≥97%
2-Acetyl-4-methylthiophene
Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 108-109 °C
Density: 1.224 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00027395
CAS Number: 13679-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ef1948301a3ca694f2a1d921b1f82dc33a63ef&vid=23ef1948301a3ca694f2a1d921b1f82dc33a63ef&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR