2-Acetyl-3-methylthiophene ≥97%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 1-(3-Methyl-2-thienyl)-1-ethanone, 1-(3-Methylthiophen-2-yl)ethanone

A167103-1G A167103-5G A167103-25G
ALA167103-1GEA 16.06 USD
ALA167103-1G ALA167103-5G ALA167103-25G
2-Acetyl-3-methylthiophene ≥97%
2-Acetyl-3-methylthiophene
Formula: C₇H₈OS
Boiling Pt: 98-99 °C
Density: 1.124 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005443
CAS Number: 13679-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7eb10c4e321ce21192209d7c8e1030b46198ea4d&vid=7eb10c4e321ce21192209d7c8e1030b46198ea4d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR