2-Azetidin-3-yl-propan-2-ol hydrochloride ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-(Azetidin-3-yl)propan-2-ol hydrochloride

A167077-1G A167077-250MG
ALA167077-1GEA 173.76 USD
ALA167077-1G ALA167077250MG
2-Azetidin-3-yl-propan-2-ol hydrochloride ≥96%
2-Azetidin-3-yl-propan-2-ol hydrochloride
Formula: C₆H₁₄ClNO
MW: 151.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22199153
CAS Number: 1357923-33-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f46908b71eae870bf49b79c44387bbeec726f19&vid=6f46908b71eae870bf49b79c44387bbeec726f19&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR