2'-Hydroxy-4'-methylacetanilide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: N-(2-Hydroxy-4-methylphenyl)acetamide

A167031-1G A167031-250MG
ALA167031-1GEA 180.41 USD
ALA167031-1G ALA167031250MG
2'-Hydroxy-4'-methylacetanilide ≥97%
2'-Hydroxy-4'-methylacetanilide
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 167-172 °C
Storage Temperature: Ambient
MDL Number: MFCD00012061
CAS Number: 13429-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f26c09e824da2256c363d35f70701bb49577dd00&vid=f26c09e824da2256c363d35f70701bb49577dd00&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR