2-Aminobenzhydrol 97

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (2-Aminophenyl)(phenyl)methanol

A166953-1G A166953-250MG A166953-25G A166953-5G
ALA166953-1GEA 15.11 USD
ALA166953-1G ALA166953250MG ALA166953-25G ALA166953-5G
2-Aminobenzhydrol 97
2-Aminobenzhydrol
Formula: C₁₃H₁₃NO
MW: 199.2500 g/mol
Melting Pt: 113-118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017097
CAS Number: 13209-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8b5fee17c777b4eee39f3b18602e7029dd99bc7&vid=b8b5fee17c777b4eee39f3b18602e7029dd99bc7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR