Azido-PEG4-C2-carboxylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethoxy]propanoic acid, Azido-PEG4-C2-acid, 1-Azido-3,6,9,12-tetraoxapentadecan-15-oic acid, Azido-PEG4-C2-carboxylic Acid

A166740-100MG A166740-1G A166740-250MG A166740-5G
ALA166740100MGEA 191.81 USD
ALA166740100MG ALA166740-1G ALA166740250MG ALA166740-5G
Azido-PEG4-C2-carboxylic acid ≥97%
Azido-PEG4-C2-carboxylic acid
Formula: C₁₁H₂₁N₃O₆
MW: 291.30 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12406155
CAS Number: 1257063-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f3c0a0503b9be383855ce8633e6b268f70f6189&vid=0f3c0a0503b9be383855ce8633e6b268f70f6189&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR