3-Allyl-1,5-naphthyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A166710-1G A166710-250MG
ALA166710-1GEA 723.81 USD
ALA166710-1G ALA166710250MG
3-Allyl-1,5-naphthyridine ≥98%
3-Allyl-1,5-naphthyridine
Formula: C₁₁H₁₀N₂
MW: 170.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17171343
CAS Number: 1246088-66-3
UN: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a23c3fd3151f6adb305d307459b5c4bde04f589&vid=6a23c3fd3151f6adb305d307459b5c4bde04f589&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR