4-Amino-7-chloroquinoline ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 7-Chloroquinolin-4-amine

A166386-1G A166386-250MG A166386-5G
ALA166386-1GEA 233.61 USD
ALA166386-1G ALA166386250MG ALA166386-5G
4-Amino-7-chloroquinoline ≥97%
4-Amino-7-chloroquinoline
Formula: C₉H₇ClN₂
MW: 178.6183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00828822
CAS Number: 1198-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ebf25579575d7d3c0f5d3a74c83c734e11fdb5e&vid=4ebf25579575d7d3c0f5d3a74c83c734e11fdb5e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR