2-Aminopyridine-4-boronic acid pinacol ester ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Aminopyridine-4-boronic acid, pinacol ester, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A166353-100MG A166353-1G A166353-250MG A166353-5G
ALA166353100MGEA 20.81 USD
ALA166353100MG ALA166353-1G ALA166353250MG ALA166353-5G
2-Aminopyridine-4-boronic acid pinacol ester ≥97%
2-Aminopyridine-4-boronic acid pinacol ester

Application:Useful boronic acid pinacol ester for synthesis of various bioactive small molecules.

Formula: C₁₁H₁₇BN₂O₂
MW: 220.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09607735
CAS Number: 1195995-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebe06a2da877042f5265eba8d1aef274359a7ed&vid=aebe06a2da877042f5265eba8d1aef274359a7ed&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR