2-Amino-4,6-difluorobenzothiazole ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A166340-1G
A166340-25G
A166340-5G
ALA166340-1GEA
17.96
USD
ALA166340-1G
ALA166340-25G
ALA166340-5G
2-Amino-4,6-difluorobenzothiazole ≥97%
2-Amino-4,6-difluorobenzothiazole
Formula:
C₇H₄F₂N₂S Melting Pt: 216-220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00276099 CAS Number: 119256-40-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...