(1-Aminocyclopropyl)methanol hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Amino-1-(hydroxymethyl)cyclopropane hydrochloride, (1-Amino-cyclopropyl)-methanol hydrochloride

A166142-1G A166142-250MG A166142-25G A166142-5G
ALA166142-1GEA 27.46 USD
ALA166142-1G ALA166142250MG ALA166142-25G ALA166142-5G
(1-Aminocyclopropyl)methanol hydrochloride ≥98%
(1-Aminocyclopropyl)methanol hydrochloride
Formula: C₄H₁₀ClNO
MW: 123.5813 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834330
CAS Number: 115652-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa2e87ef7cb4204ed8498c157a3757d76c926366&vid=aa2e87ef7cb4204ed8498c157a3757d76c926366&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR