3-(4-Chlorophenyl)glutaramic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A166064-1G A166064-25G A166064-5G
ALA166064-1GEA 14.16 USD
ALA166064-1G ALA166064-25G ALA166064-5G
3-(4-Chlorophenyl)glutaramic acid ≥98%
3-(4-Chlorophenyl)glutaramic acid
Formula: C₁₁H₁₂ClNO₃
Melting Pt: 168-170°C
Storage Temperature: Ambient
MDL Number: MFCD00190251
CAS Number: 1141-23-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2eff9011940fe0cba5d728008eec00eac5e9064e&vid=2eff9011940fe0cba5d728008eec00eac5e9064e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR