5-Amino-2,4-difluorophenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A166012-100MG A166012-1G A166012-250MG A166012-25G A166012-50MG A166012-5G
ALA166012100MGEA 79.71 USD
ALA166012100MG ALA166012-1G ALA166012250MG ALA166012-25G ALA166012-50MG ALA166012-5G
5-Amino-2,4-difluorophenol ≥98%
5-Amino-2,4-difluorophenol
Formula: C₆H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD04973718
CAS Number: 113512-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09828f8c23a2704ae9c9f754fbb097ae32fbddbd&vid=09828f8c23a2704ae9c9f754fbb097ae32fbddbd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR