3-Amino-1,2,4-triazine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A165953-100G A165953-10G A165953-1G A165953-50G A165953-5G
ALA165953-100GEA 756.11 USD
ALA165953-100G ALA165953-10G ALA165953-1G ALA165953-50G ALA165953-5G
3-Amino-1,2,4-triazine ≥97%
3-Amino-1,2,4-triazine
Formula: C₃H₄N₄
Melting Pt: 175.5°C
Storage Temperature: Ambient
MDL Number: MFCD00006459
CAS Number: 1120-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fcde093fbcab61744e1649e0af2e0477c19f5ce&vid=2fcde093fbcab61744e1649e0af2e0477c19f5ce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR