((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
Adefovir
A165714-100G
A165714-1G
A165714-5G
ALA165714-100GEA
175.66
USD
ALA165714-100G
ALA165714-1G
ALA165714-5G
((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid ≥98% (by HPLC)
((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid
Adefovir (GS-0393) is an adenosine monophosphate analog antiviral agent that after intracellular conversion to Adefovir diphosphate inhibits HBV DNA polymerase. Adefovir has an IC50 of 0.7 μM against HBV in the HepG2.2.15 cell line. Adefovir has good antiviral activity against several viruses, including HBV and herpesviruses.
Formula:
C₈H₁₂N₅O₄P MW: 273.19 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00866943 CAS Number: 106941-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...