3-Amino-5-methoxyisonicotinonitrile ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

A165620-1G A165620-250MG A165620-50MG
ALA165620-1GEA 575.61 USD
ALA165620-1G ALA165620250MG ALA165620-50MG
3-Amino-5-methoxyisonicotinonitrile ≥95%
3-Amino-5-methoxyisonicotinonitrile
Formula: C₇H₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD11052851
CAS Number: 1045855-60-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cc01b53fc6c9553591617ebd2bf00047b645620&vid=3cc01b53fc6c9553591617ebd2bf00047b645620&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR