4-Amino-6-methoxy-2-methylquinoline ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A165608-1G A165608-250MG
ALA165608-1GEA 201.31 USD
ALA165608-1G ALA165608250MG
4-Amino-6-methoxy-2-methylquinoline ≥97%
4-Amino-6-methoxy-2-methylquinoline
Formula: C₁₁H₁₂N₂O
MW: 188.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00552889
CAS Number: 104217-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70ea8452a542b70bb83d27602b084f6cfe384ff5&vid=70ea8452a542b70bb83d27602b084f6cfe384ff5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR