(4-Aminophenylthio)acetic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A165606-1G A165606-25G A165606-5G
ALA165606-1GEA 45.51 USD
ALA165606-1G ALA165606-25G ALA165606-5G
(4-Aminophenylthio)acetic acid ≥97%
(4-Aminophenylthio)acetic acid
Formula: C₈H₉NO₂S
Melting Pt: 200°C
Storage Temperature: Ambient
MDL Number: MFCD00025360
CAS Number: 104-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1be5a3819824c83c4198dc77dc9cdcaebe158f1&vid=a1be5a3819824c83c4198dc77dc9cdcaebe158f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR